Pyrazine-2-carbothioic acid, S-phenyl ester ≥97%

Supplier: Matrix Scientific

007388-500MG
101792-630EA 144.62 USD
101792-630
Pyrazine-2-carbothioic acid, S-phenyl ester ≥97%
Pyrazine-2-carbothioic acid, S-phenyl ester

Hazard: Irritant/Refrigerate

Formula: C₁₁H₈N₂OS
MW: 216.26 g/mol
Melting Pt: 149…151 °C
MDL Number: MFCD03425980
CAS Number: 121761-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR