3-(Dimethylcarbamoyl)phenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Dimethylcarbamoyl)benzeneboronic acid(3-(Dimethylcarbamoyl)phenyl)boronic acid3-(N,N-Dimethylaminocarbonyl)phenylboronic acid

007380-500MG
101792-602EA 23.08 USD
101792-602
3-(Dimethylcarbamoyl)phenylboronic acid ≥97%
3-(Dimethylcarbamoyl)phenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₉H₁₂BNO₃
MW: 193.01 g/mol
MDL Number: MFCD03412066
CAS Number: 373384-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR