Bis(hexylene glycolato)diboron ≥97%
Supplier: Matrix Scientific
Synonyms:
4,4,4',4',6,6'-Hexamethyl-2,2'-bi(1,3,2-dioxaborinane)
, 4,4,4',4',6,6'-Hexamethyl-2,2'-bi-1,3,2-dioxaborinane
007368-5G
101792-572EA
73.85
USD
101792-572
Bis(hexylene glycolato)diboron ≥97%
Bis(hexylene glycolato)diboron
Purity: 97%
CAS: 230299-21-5
MDL Number: MFCD06246008
Molecular Formula: C12H24B2O4
Molecular Weight: 253,94
Hazard: Irritant
Formula:
C₁₂H₂₄B₂O₄ MW: 253.94 g/mol |
MDL Number:
MFCD06246008 CAS Number: 230299-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...