2-(Diphenylphosphino)-2'-(N,N-dimethylamino)biphenyl ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Diphenylphosphino-2'-(N,N-dimethylamino))-1,1'-biphenylPhDavePhos2-Diphenylphosphino-2'-(N,N-dimethylamino)biphenyl

007254-1G
101792-260EA 224.62 USD
101792-260
2-(Diphenylphosphino)-2'-(N,N-dimethylamino)biphenyl ≥98%
2-(Diphenylphosphino)-2'-(N,N-dimethylamino)biphenyl

Hazard: Irritant

Formula: C₂₆H₂₄NP
MW: 381.46 g/mol
Melting Pt: 124…126 °C
MDL Number: MFCD03094578
CAS Number: 240417-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR