1-[4-(Trifluoromethyl)phenyl]-2-pyrrolidinone

Supplier: Matrix Scientific

007042-1G
101791-888EA 56.92 USD
101791-888
1-[4-(Trifluoromethyl)phenyl]-2-pyrrolidinone
1-[4-(Trifluoromethyl)phenyl]-2-pyrrolidinone

Hazard: Irritant

Formula: C₁₁H₁₀F₃NO
MDL Number: MFCD00138491
CAS Number: 73081-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR