5-(4-Nitrophenyl)isoxazole
Supplier: Matrix Scientific
007033-1G
101791-878EA
283.08
USD
101791-878
5-(4-Nitrophenyl)isoxazole
5-(4-Nitrophenyl)isoxazole
Hazard: Irritant
Formula:
C₉H₆N₂O₃ Melting Pt: 176…178 °C |
CAS Number:
3383-42-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...