5-(4-Fluorophenyl)isoxazole

Supplier: Matrix Scientific

007027-1G
101791-862EA 15.38 USD
101791-862
5-(4-Fluorophenyl)isoxazole
5-(4-Fluorophenyl)isoxazole

Hazard: Irritant

Formula: C₉H₆FNO
Melting Pt: 51…53 °C
MDL Number: MFCD02183544
CAS Number: 138716-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR