5-(3,4-Difluorophenyl)isoxazole

Supplier: Matrix Scientific

007021-1G
101791-844EA 203.08 USD
101791-844
5-(3,4-Difluorophenyl)isoxazole
5-(3,4-Difluorophenyl)isoxazole

Hazard: Irritant

Formula: C₉H₅F₂NO
MDL Number: MFCD02183541
CAS Number: 387358-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR