5-(2,6-Difluorophenyl)isoxazole

Supplier: Matrix Scientific

007020-1G
101791-842EA 148.35 USD
101791-842
5-(2,6-Difluorophenyl)isoxazole
5-(2,6-Difluorophenyl)isoxazole

Hazard: Irritant

Formula: C₉H₅F₂NO
MDL Number: MFCD02183540
CAS Number: 138716-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR