5-(2,4-Difluorophenyl)isoxazole

Supplier: Matrix Scientific

007018-1G
101791-838EA 123.08 USD
101791-838
5-(2,4-Difluorophenyl)isoxazole
5-(2,4-Difluorophenyl)isoxazole

Hazard: Irritant

Formula: C₉H₅F₂NO
MDL Number: MFCD02183538
CAS Number: 138716-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR