2,3-Difluorobenzhydrazide

Supplier: Matrix Scientific

007014-1G
101791-828EA 169.23 USD
101791-828
2,3-Difluorobenzhydrazide
2,3-Difluorobenzhydrazide

Hazard: Irritant

Formula: C₇H₆F₂N₂O
MDL Number: MFCD02183536
CAS Number: 387358-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR