5-(3-Bromophenyl)isoxazole

Supplier: Matrix Scientific

006999-500MG
101791-786EA 38.46 USD
101791-786
5-(3-Bromophenyl)isoxazole
5-(3-Bromophenyl)isoxazole

Hazard: Irritant

Formula: C₉H₆BrNO
MW: 224.06 g/mol
MDL Number: MFCD02183531
CAS Number: 7064-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR