4-(Methoxycarbonyl)phenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Methoxycarbonyl)benzeneboronic acid(4-(Methoxycarbonyl)phenyl)boronic acid

006785-25G
101791-106EA 83.08 USD
101791-106
4-(Methoxycarbonyl)phenylboronic acid ≥97%
4-(Methoxycarbonyl)phenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₈H₉BO₄
MW: 179.97 g/mol
Melting Pt: 220…225 °C
MDL Number: MFCD01632203
CAS Number: 99768-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR