4-(N,N-Dimethylamino)phenylboronic acid hydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(N,N-Dimethylamino)Phenylboronic Acid Hydrochloride

006774-1G 006774-5G
101791-076EA 44.62 USD
101791-076 101791-078
4-(N,N-Dimethylamino)phenylboronic acid hydrochloride ≥97%
4-(N,N-Dimethylamino)phenylboronic acid hydrochloride

Hazard: Irritant/Air Sensitive/Refrigerate

Formula: C₈H₁₃BClNO₂
MW: 201.4583 g/mol
MDL Number: MFCD04112547
CAS Number: 1150114-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR