4-Borono-DL-phenylalanine ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(2-Amino-2-carboxyethyl)benzeneboronic acid

006756-500MG
101791-026EA 153.85 USD
101791-026
4-Borono-DL-phenylalanine ≥97%
4-Borono-DL-phenylalanine

Hazard: Irritant

Formula: C₉H₁₂BNO₄
MW: 209.01 g/mol
MDL Number: MFCD01863704
CAS Number: 90580-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR