Cyclopentylboronic acid ≥97%

Supplier: Matrix Scientific

006629-25G 006629-5G
102550-876EA 206.15 USD
102550-876 101790-846
Cyclopentylboronic acid ≥97%
Cyclopentylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₅H₁₁BO₂
MW: 113.95 g/mol
Melting Pt: 108…109 °C
MDL Number: MFCD01074541
CAS Number: 63076-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR