N,N-Dibutylbenzenesulfonamide ≥98%

Supplier: Matrix Scientific

006627-1G 006627-5G
101790-836EA 30.77 USD
101790-836 101790-838
N,N-Dibutylbenzenesulfonamide ≥98%
N,N-Dibutylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₂₃NO₂S
Melting Pt: 211 °C
MDL Number: MFCD00151808
CAS Number: 5339-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR