Ethyl-α-cyano-2-nitro-4-(trifluoromethyl)phenylacetate ≥97%

Supplier: Matrix Scientific

006577-1G 006577-5G
101790-788EA 76.92 USD
101790-788 101790-790
Ethyl-α-cyano-2-nitro-4-(trifluoromethyl)phenylacetate ≥97%
Ethyl-alpha-cyano-2-nitro-4-(trifluoromethyl)phenylacetate

Hazard: Irritant

Formula: C₁₂H₉F₃N₂O₄
MW: 302.21 g/mol
Melting Pt: 61…62 °C
MDL Number: MFCD01861144
CAS Number: 13544-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR