Boc-L-pentafluorophenylalanine

Supplier: Matrix Scientific

Synonyms: Boc-pentafluoro-Phe-OH

006268-1G
101790-306EA 207.69 USD
101790-306
Boc-L-pentafluorophenylalanine
Boc-L-pentafluorophenylalanine

CAS: 34702-60-8
MDL Number: MFCD00151893
Molecular Formula: C14H14F5NO4
Molecular Weight: 355,26
Hazard: Irritant/Refrigerate

Formula: C₁₄H₁₄F₅NO₄
MW: 355.261 g/mol
Melting Pt: 106…108 °C
MDL Number: MFCD00151893
CAS Number: 34702-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR