3-(Difluoromethoxy)benzylamine

Supplier: Matrix Scientific

Synonyms: (3-(Difluoromethoxy)phenyl)methanamine

006006-500MG
101789-904EA 77.05 USD
101789-904
3-(Difluoromethoxy)benzylamine
3-(Difluoromethoxy)benzylamine

Hazard: Irritant

Formula: C₈H₉F₂NO
MW: 173.16 g/mol
MDL Number: MFCD00236230
CAS Number: 244022-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR