4-(Trifluoromethyl)-DL-phenylalanine
Supplier: Matrix Scientific
005414-500MG
101788-902EA
95.38
USD
101788-902
4-(Trifluoromethyl)-DL-phenylalanine
4-(Trifluoromethyl)-DL-phenylalanine
Hazard: Irritant-Harmful
Formula:
C₁₀H₁₀F₃NO₂ |
MDL Number:
MFCD00070813 CAS Number: 14091-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...