Ethyl-6-bromo-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥96%
Supplier: Matrix Scientific
070398-5G
070398-1G
070398-500MG
101944-576EA
1106.15
USD
101944-576
101944-580
101944-578
Ethyl-6-bromo-1H-pyrrolo[2,3-b]pyridine-2-carboxylate ≥96%
Ethyl-6-bromo-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₉BrN₂O₂ |
MDL Number:
MFCD11518923 CAS Number: 577711-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...