4-(Trimethylsilyl)phenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (4-(Trimethylsilyl)phenyl)boronic acid

070397-5G
101944-572EA 136.92 USD
101944-572
4-(Trimethylsilyl)phenylboronic acid ≥97%
4-(Trimethylsilyl)phenylboronic acid

Hazard: Irritant/Refrigerate

Formula: C₉H₁₅BO₂Si
MW: 194.1100 g/mol
MDL Number: MFCD00093348
CAS Number: 17865-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR