5-Methoxyisoindoline ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methoxyisoindoline hydrochloride

070341-5G 070341-1G
101944-330EA 6975.38 USD
101944-330 101944-328
5-Methoxyisoindoline ≥95%
5-Methoxyisoindoline

Hazard: Irritant

Formula: C₉H₁₁NO
MW: 149.19 g/mol
MDL Number: MFCD06804552
CAS Number: 127168-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR