Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloroimidazo[4,5-c]pyridine ≥95%

Supplier: Matrix Scientific


070335-1G 070335-5G
101944-304EA 549.23 USD InStock
101944-304 101944-306
4-Chloroimidazo[4,5-c]pyridine ≥95%
4-Chloroimidazo[4,5-c]pyridine
Purity: 95%
CAS: 2770-01-6
MDL Number: MFCD08689582
Molecular Formula: C6H4ClN3
Molecular Weight: 153,57
Hazard: Irritant


Formula: C₆H₄ClN₃
Melting Pt: 179…181 °C
MDL Number: MFCD08689582
CAS Number: 2770-01-6
EINECS: 000-000-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty