2-(Cyclopropylmethoxy)-5-methylphenylboronic acid pinacol ester ≥98%

Supplier: Matrix Scientific

070326-1G 070326-500MG
101944-268EA 524.62 USD
101944-268 101944-270
2-(Cyclopropylmethoxy)-5-methylphenylboronic acid pinacol ester ≥98%
2-(Cyclopropylmethoxy)-5-methylphenylboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₇H₂₅BO₃
MDL Number: MFCD13152008
CAS Number: 1185836-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR