3-Bromo-1H-pyrazolo[4,3-b]pyridine ≥96%
Supplier: Matrix Scientific
070314-500MG
070314-5G
070314-1G
101944-214EA
930.77
USD
101944-214
101944-218
101944-216
3-Bromo-1H-pyrazolo[4,3-b]pyridine ≥96%
3-Bromo-1H-pyrazolo[4,3-b]pyridine
Hazard: Irritant
Formula:
C₆H₄BrN₃ MW: 198.0201 g/mol |
MDL Number:
MFCD11518974 CAS Number: 633328-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...