2-Methyl-5-thiazolecarboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methylthiazole-5-carboxylic acid

070312-5G
101944-208EA 52.31 USD
101944-208
2-Methyl-5-thiazolecarboxylic acid ≥97%
2-Methyl-5-thiazolecarboxylic acid

Hazard: Irritant/Refrigerate

Formula: C₅H₅NO₂S
MW: 143.1637 g/mol
MDL Number: MFCD01760300
CAS Number: 40004-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR