(S)-(-)-Indoline-2-carboxylic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
(S)-Indoline-2-carboxylic acid
070294-25G
101944-130EA
223.08
USD
101944-130
(S)-(-)-Indoline-2-carboxylic acid ≥98%
(S)-(-)-Indoline-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₉NO₂ MW: 163.18 g/mol Melting Pt: 178…180 °C |
MDL Number:
MFCD00070578 CAS Number: 79815-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...