Ethyl 5-aminoimidazo[1,2-a]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

070287-1G 070287-5G
101944-104EA 1396.92 USD
101944-104 101944-106
Ethyl 5-aminoimidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 5-aminoimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₂
MDL Number: MFCD09842618
CAS Number: 1000017-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR