Methyl-2-(3-nitro-2-oxopyridin-1(2H)-yl)acetate ≥95%

Supplier: Matrix Scientific

070285-1G 070285-5G
101944-098EA 5273.85 USD
101944-098 101944-096
Methyl-2-(3-nitro-2-oxopyridin-1(2H)-yl)acetate ≥95%
Methyl-2-(3-nitro-2-oxopyridin-1(2H)-yl)acetate

Hazard: Irritant

Formula: C₈H₈N₂O₅
MDL Number: MFCD11846205
CAS Number: 183666-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR