7-(Trifluoromethyl)-1,2,3,4-tetrahydro-5-iodoisoquinoline hydrochloride ≥95%

Supplier: Matrix Scientific

070266-1G 070266-5G
101944-022EA 6455.38 USD
101944-022 101944-020
7-(Trifluoromethyl)-1,2,3,4-tetrahydro-5-iodoisoquinoline hydrochloride ≥95%
7-(Trifluoromethyl)-1,2,3,4-tetrahydro-5-iodoisoquinoline hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₀ClF₃IN
MDL Number: MFCD11846196
CAS Number: 1187830-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR