Pyrrolo[1,2-a]pyrazin-1(2H)-one ≥95%
Supplier: Matrix Scientific
070263-5G
070263-1G
101944-010EA
5438.46
USD
101944-010
101944-008
Pyrrolo[1,2-a]pyrazin-1(2H)-one ≥95%
Pyrrolo[1,2-a]pyrazin-1(2H)-one
Hazard: Irritant
Formula:
C₇H₆N₂O |
MDL Number:
MFCD09834911 CAS Number: 136927-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...