7-Chlorothieno[3,2-b]pyridine oxalic acid ≥95%

Supplier: Matrix Scientific

070259-1G 070259-5G
101943-992EA 1396.92 USD
101943-992 101943-994
7-Chlorothieno[3,2-b]pyridine oxalic acid ≥95%
7-Chlorothieno[3,2-b]pyridine oxalic acid

Hazard: Irritant

Formula: C₉H₆ClNO₄S
MDL Number: MFCD11846193
CAS Number: 1187830-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR