7-Bromofuro[3,2-c]pyridin-4(5H)-one ≥95%

Supplier: Matrix Scientific

070254-1G 070254-5G
101943-972EA 1716.92 USD
101943-972 101943-974
7-Bromofuro[3,2-c]pyridin-4(5H)-one ≥95%
7-Bromofuro[3,2-c]pyridin-4(5H)-one

Hazard: Irritant

Formula: C₇H₄BrNO₂
MDL Number: MFCD11846190
CAS Number: 603301-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR