Ethyl-2-(3-chloro-2-oxopyrazin-1(2H)-yl)acetate ≥95%

Supplier: Matrix Scientific

070242-1G 070242-5G
101943-924EA 1396.92 USD
101943-924 101943-926
Ethyl-2-(3-chloro-2-oxopyrazin-1(2H)-yl)acetate ≥95%
Ethyl-2-(3-chloro-2-oxopyrazin-1(2H)-yl)acetate

Hazard: Irritant

Formula: C₈H₉ClN₂O₃
MDL Number: MFCD11973832
CAS Number: 1194374-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR