5-Bromo-4-chloro-1H-indazole ≥97%

Supplier: Matrix Scientific

070179-1G 070179-5G
101943-682EA 553.85 USD
101943-682 101943-684
5-Bromo-4-chloro-1H-indazole ≥97%
5-Bromo-4-chloro-1H-indazole

Hazard: Irritant

Formula: C₇H₄BrClN₂
MW: 231.4771 g/mol
MDL Number: MFCD11007933
CAS Number: 1082041-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR