5-Bromo-7-chloro-1H-indazole ≥97%

Supplier: Matrix Scientific

070178-1G 070178-5G
101943-678EA 930.77 USD
101943-678 101943-680
5-Bromo-7-chloro-1H-indazole ≥97%
5-Bromo-7-chloro-1H-indazole

Hazard: Irritant

Formula: C₇H₄BrClN₂
MW: 231.4771 g/mol
MDL Number: MFCD11044588
CAS Number: 635712-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR