2-Chloro-3-fluoro-5-(tributylstannyl)pyridine ≥94%
Supplier: Matrix Scientific
070175-1G
070175-500MG
070175-5G
101943-662EA
640
USD
101943-662
101943-664
101943-666
2-Chloro-3-fluoro-5-(tributylstannyl)pyridine ≥94%
2-Chloro-3-fluoro-5-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₇H₂₉ClFNSn |
MDL Number:
MFCD12827556 CAS Number: 405556-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...