2-Amino-4-chloro-3-nitropyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-3-nitropyridin-2-amine

070104-5G
102833-474EA 50.6 USD
102833-474
2-Amino-4-chloro-3-nitropyridine ≥95%
2-Amino-4-chloro-3-nitropyridine

Hazard: Irritant

Formula: C₅H₄ClN₃O₂
MW: 173.56 g/mol
Melting Pt: 182…184 °C
MDL Number: MFCD04116097
CAS Number: 6980-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR