5-Bromoimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

070102-1G
101943-334EA 233.85 USD
101943-334
5-Bromoimidazo[1,2-a]pyridine ≥97%
5-Bromoimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
MDL Number: MFCD08275698
CAS Number: 69214-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR