2-Bromo-6-(tributylstannyl)pyridine

Supplier: Matrix Scientific

070098-1G
101943-316EA 524.62 USD
101943-316
2-Bromo-6-(tributylstannyl)pyridine
2-Bromo-6-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₇H₃₀BrNSn
CAS Number: 189083-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR