5-Bromo-1,7-dimethyl-1H-indazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-1,7-dimethylindazole

070080-5G
101943-234EA 2416.92 USD
101943-234
5-Bromo-1,7-dimethyl-1H-indazole ≥95%
5-Bromo-1,7-dimethyl-1H-indazole

Hazard: Irritant

Formula: C₉H₉BrN₂
MW: 225.09 g/mol
MDL Number: MFCD11226595
CAS Number: 1092352-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR