Isobutyramide ≥98%

Supplier: Matrix Scientific

070073-1KG 070073-25G
101943-194EA 146.15 USD
101943-194 101943-198
Isobutyramide ≥98%
Isobutyramide

Hazard: Irritant

Formula: C₄H₉NO
MW: 87.12 g/mol
MDL Number: MFCD00008019
CAS Number: 563-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR