2,5-Dibromothieno[3,2-b]thiophene ≥98%

Supplier: Matrix Scientific

070070-1G
101943-182EA 198.46 USD
101943-182
2,5-Dibromothieno[3,2-b]thiophene ≥98%
2,5-Dibromothieno[3,2-b]thiophene

Hazard: Toxic/Store in Freezer

Formula: C₆H₂Br₂S₂
MW: 298.0181 g/mol
Melting Pt: 124…129 °C
MDL Number: MFCD03931293
CAS Number: 25121-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR