1H-Indazole-6-boronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Indazolyboronic acid1H-Indazol-6-yl-6-boronic acid1H-Indazol-6-ylboronic acid

070067-1G
101943-168EA 56.92 USD
101943-168
1H-Indazole-6-boronic acid ≥97%
1H-Indazole-6-boronic acid

Hazard: Irritant

Formula: C₇H₇BN₂O₂
MW: 161.9500 g/mol
MDL Number: MFCD04972419
CAS Number: 885068-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR