Methyl-3-cyanoindole-6-carboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 3-cyano-1H-indole-6-carboxylate

070064-1G
101943-150EA 815.38 USD
101943-150
Methyl-3-cyanoindole-6-carboxylate ≥98%
Methyl-3-cyanoindole-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₈N₂O₂
MW: 200.197 g/mol
MDL Number: MFCD09878564
CAS Number: 1000576-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR