2-Amino-5-chloro-4-methylpyridine ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Amino-5-chloro-4-picoline
070059-5G
101943-134EA
743.08
USD
101943-134
2-Amino-5-chloro-4-methylpyridine ≥98%
2-Amino-5-chloro-4-methylpyridine
Hazard: Irritant
Formula:
C₆H₇ClN₂ MW: 142.5900 g/mol |
MDL Number:
MFCD06410759 CAS Number: 36936-27-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...