6-(Tributylstannyl)[1,2,4]triazolo[1,5-a]pyridine ≥95%

Supplier: Matrix Scientific

070052-1G
101943-100EA 1396.92 USD
101943-100
6-(Tributylstannyl)[1,2,4]triazolo[1,5-a]pyridine ≥95%
6-(Tributylstannyl)[1,2,4]triazolo[1,5-a]pyridine

Hazard: Irritant

Formula: C₁₈H₃₁N₃Sn
MDL Number: MFCD13151992
CAS Number: 1204580-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR