(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (2S)-1-[(tert-butoxy)carbonyl]-4,4-difluoropyrrolidine-2-carboxylic acidN-(tert-Butoxycarbonyl)-4,4-difluoro-L-prolineN-Boc-4,4-difluoro-L-proline(S)-1-Boc-4,4-difluoro-2-pyrrolidinecarboxylic Acid(S)-1-(tert-Butoxycarbonyl)-4,4-difluoropyrrolidine-2-carboxylic acid

070034-5G
101943-028EA 384.62 USD
101943-028
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid ≥97%
(S)-1-(tert-Butoxycarbonyl)-4,4-difluoro-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₅F₂NO₄
MW: 251.23 g/mol
MDL Number: MFCD03094917
CAS Number: 203866-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR